Anti-Helicobacter pylori Mouse Monoclonal Antibody (CF405S) [clone: HP/212]

Supplier: Biotium
BNC040212-500 BNC040212-100
BNC040212-500EA 560 EUR
BNC040212-500 BNC040212-100
Anti-Helicobacter pylori Mouse Monoclonal Antibody (CF405S) [clone: HP/212]
Antibodies
The spiral shaped bacteriumHelicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy.H. pyloriis known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach.This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach.

Type: Primary
Antigen: Helicobacter pylori
Clonality: Monoclonal
Clone: HP/212
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Helicobacter pylori
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR