Anti-ZAP70 Mouse Monoclonal Antibody (Purified) [clone: ZAP70/528]
Supplier: Biotium
BNUB0528-500 BNUB0528-100
BNUB0528-500EA 511 EUR
BNUB0528-500 BNUB0528-100
Anti-ZAP70 Mouse Monoclonal Antibody (Purified) [clone: ZAP70/528]
Antibodies
ZAP70 is a 70 kDa protein tyrosine kinase found in T-cells and natural killer cells.Control of this protein translation is via the IgVH gene. ZAP70 protein is expressed in leukemic cells of approximately 25% of chronic lymphocytic leukemia (CLL) cases as well.Anti-ZAP70 expression is an excellent surrogate marker for the distinction between the Ig-mutated (anti-ZAP70 negative) and Ig-unmutated (anti-ZAP70 positive) CLL subtypes and can identify patient groups with divergent clinical courses. The anti-ZAP70 positive Ig-unmutated CLL cases have been shown to have a poorer prognosis.
Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: ZAP70/528
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human