Anti-KDR Mouse Monoclonal Antibody (CF488A) [clone: KDR657]

Supplier: Biotium
BNC880657-500 BNC880657-100
BNC880657-500EA 560 EUR
BNC880657-500 BNC880657-100
Anti-KDR Mouse Monoclonal Antibody (CF488A) [clone: KDR657]
Antibodies
CD309, also known as VEGF-R2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGF-R2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGF-R2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGF-R2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cellMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KDR
Antigen name VEGFR2 / Flk-1 / KDR3 / CD309
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone Clone KDR657
Reactivity Human
Host Mouse
Gene ID 479756
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR