Anti-MBP Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-2967REA 183 EUR
BOSSBS-2967R
Anti-MBP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR