Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-3016REA 710 EUR
BOSSBS-3016R
Anti-MAPK3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR