Anti-KDR Rat Monoclonal Antibody (CF647) [clone: KDR721]

Supplier: Biotium
BNC470721-500 BNC470721-100
BNC470721-500EA 560 EUR
BNC470721-500 BNC470721-100
Anti-KDR Rat Monoclonal Antibody (CF647) [clone: KDR721]
Antibodies
CD309, also known as VEGFR2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGFR2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGFR2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGFR2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cellMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KDR KDR
Antigen name VEGFR2 / Flk-1 / KDR3 / CD309 VEGFR2 / Flk-1 / KDR3 / CD309
Environmentally Preferable
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation CF® 647 CF® 647
Clone Clone KDR721 Clone KDR721
Reactivity Human Human
Host Rat Rat
Gene ID 479756 479756
Isotype IgG1 kappa IgG1 kappa
ImmunoFluorescence Yes
Concentration 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR