Anti-PRLR Mouse Monoclonal Antibody [clone: B6.2 PRLR742]

Supplier: Biotium
BNUB0744-500 BNUB0744-100
BNUB0744-500EA 511 EUR
BNUB0744-500 BNUB0744-100
Anti-PRLR Mouse Monoclonal Antibody [clone: B6.2 PRLR742]
Antibodies
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

  • Purified

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: B6.2 PRLR742
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR