Anti-FUT3 Mouse Monoclonal Antibody (Purified) [clone: 2-25LE]

Supplier: Biotium
BNUB1008-500 BNUB1008-100
BNUB1008-500EA 511 EUR
BNUB1008-500 BNUB1008-100
Anti-FUT3 Mouse Monoclonal Antibody (Purified) [clone: 2-25LE]
Antibodies
The Lewis histo-blood group system comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This gene is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol FUT3
Antigen name Lewis B
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone 2-25LE
Reactivity Pig,Human,Guinea pig
Host Mouse
Gene ID 77961
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR