Anti-PAX7 Mouse Monoclonal Antibody (CF594) [clone: PAX7/1187]

Supplier: Biotium
BNC941187-500 BNC941187-100
BNC941187-500EA 560 EUR
BNC941187-500 BNC941187-100
Anti-PAX7 Mouse Monoclonal Antibody (CF594) [clone: PAX7/1187]
Antibodies
The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma).

Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7/1187
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR