Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD494]

Supplier: Biotium

BNUM0494-50EA 395 EUR
BNUM0494-50
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD494]
Antibodies
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD494
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR