Hydrochloric acid 2 mol/l (2 N) for biopharmaceutical production
Supplier: VWR Chemicals
|
Warning
|
85399.460EA
0
EUR
85399.460
Hydrochloric acid 2 mol/l (2 N) for biopharmaceutical production
Hydrochloric acid
Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula:
HCl MW: 36,46 g/mol Density: 1,031 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Specification Test Results
Evaporation residue | Max. 100 ppm |
Free chlorine | Max. 1 ppm |
Heavy metals (as Pb) | Max. 1 ppm |
Ignition residue (SO₄) | Max. 80 ppm |
SO₃ (Sulphite) | Max. 10 ppm |
SO₄ (Sulphate) | Max. 5 ppm |
As (Arsenic) | Max. 1.5 ppm |
Residual metal catalysts | Unlikely to be present |
Residual solvents (ICH Q3C) | Excl.by manufact. process |
Endotoxin | Max. 0.25 EU/ml |
Filtered at 0.2 µm | Confirmed |
Titer | 1.8 - 2.2 mol/l |
Appearance | Passes test |
Free bromine or chlorine | Passes test |
Identification | Passes test |
Colouration | Max. 10 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...