Bulk adsorbents, Ottawa sand
Supplier: Restek
REST26137EA 400 EUR
REST26137
Bulk adsorbents, Ottawa sand
Chromatography Sorbents SPE Sorbents
Ottawa sand is listed in several U.S. EPA methods as the specified medium for matrix spike and laboratory control blanks.
Balení: Packaged in convenient 5 kg buckets.