CarboBlack columns (packed), silco smooth tubing
Supplier: Restek
80105-800 80105-800901 80105-810 80105-820 80105-830 80105-840 80105-850 80105-853 80105-895 80105-896 80106-800 80106-810 80106-810901 80106-820 80106-830 80106-840 80106-850 80106-892 80106-894 80107-800 80107-810 80107-811 80107-820 80107-830 80107-840 80111-800 80111-810 80111-820 80111-830 80116-800 80116-810 80116-820 80116-830 80116-840 80116-852 80116-895 80116-896 80121-800 80121-810 80121-820 80121-830 80121-840 80121-840901 80122-800 80122-810 80122-810901 80122-820 80122-830 80122-840 80205-800 80205-810 80205-820 80205-830 80205-840 80205-895 80206-800 80206-800901 80206-810 80206-810901 80206-820 80206-820901 80206-830 80206-840 80206-840901 80206-845901 80206-850901 80207-800 80207-810 80207-820 80207-820901 80207-830 80207-840 80207-850 80211-800 80211-810 80211-820 80211-830 80211-840 80216-800 80216-810 80216-820 80216-830 80305-800 80305-810 80305-820 80305-830 80311-800 80311-810 80311-820 80311-830 80311-840 80105-810901 80105-811 80107-850 80107-895 80122-850 80305-840
REST80105-800EA 405 EUR
REST80105-800 REST80105-800901 REST80105-810 REST80105-820 REST80105-830 REST80105-840 REST80105-850 REST80105-853 REST80105-895 REST80105-896 REST80106-800 REST80106-810 REST80106-810901 REST80106-820 REST80106-830 REST80106-840 REST80106-850 REST80106-892 REST80106-894 REST80107-800 REST80107-810 REST80107-811 REST80107-820 REST80107-830 REST80107-840 REST80111-800 REST80111-810 REST80111-820 REST80111-830 REST80116-800 REST80116-810 REST80116-820 REST80116-830 REST80116-840 REST80116-852 REST80116-895 REST80116-896 REST80121-800 REST80121-810 REST80121-820 REST80121-830 REST80121-840 REST80121-840901 REST80122-800 REST80122-810 REST80122-810901 REST80122-820 REST80122-830 REST80122-840 REST80205-800 REST80205-810 REST80205-820 REST80205-830 REST80205-840 REST80205-895 REST80206-800 REST80206-800901 REST80206-810 REST80206-810901 REST80206-820 REST80206-820901 REST80206-830 REST80206-840 REST80206-840901 REST80206-845901 REST80206-850901 REST80207-800 REST80207-810 REST80207-820 REST80207-820901 REST80207-830 REST80207-840 REST80207-850 REST80211-800 REST80211-810 REST80211-820 REST80211-830 REST80211-840 REST80216-800 REST80216-810 REST80216-820 REST80216-830 REST80305-800 REST80305-810 REST80305-820 REST80305-830 REST80311-800 REST80311-810 REST80311-820 REST80311-830 REST80311-840 REST80105-810901 REST80105-811 REST80107-850 REST80107-895 REST80122-850 REST80305-840
CarboBlack columns (packed), silco smooth tubing
Chromatography Columns
Graphitised carbon black offers unique selectivity and very little adsorption for alcohol analyses. Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B support, with its higher surface area, can hold up to a 10% loading of a non silicone liquid phase. CarboBlack C support can hold up to a 1% loading of a non silicone liquid phase.
  • CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds