Capillary columns, Rtx®-PCB (fused silica)
Supplier: Restek
41302 41304 41303 13226 13239
REST41302EA 622 EUR
REST41302 REST41304 REST41303 REST13226 REST13239
Capillary columns, Rtx®-PCB (fused silica)
Chromatography Columns
These proprietary Crossbond phase columns with unique polymer are suitable for PCBs analysis by GC-ECD or GC-MS.
  • Good results for other semivolatiles
  • Low polarity and are inert to active compounds
  • Temperature stability up to 340 °C