Inlet liner removal tool
Supplier: Restek
REST20181EA 46 EUR
REST20181
Inlet liner removal tool
Puller and Inserter Tools
This tool is made from high-temperature silicone and it provides easy removal of liner from injector.
  • Protect against finger burns
  • It will not chip or crack the liner