NGAL (rat) ELISA kit
Supplier: Enzo Life Sciences
ALEXBPD-KIT-046EA 1270 EUR
ALEXBPD-KIT-046 ENZOBPDKIT046
NGAL (rat) ELISA kit
Assays ELISAs
For the quantitative determination of NGAL in rat urine, plasma, serum, tissue extracts, or culture supernatants.
  • Ready to use pre-coated plate and individual calibrators save time and reduce errors
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
  • Absorbance is read at 450 nm
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in rat samples. Absorbance is read at 450nm.
Delivery information: The kit contains a microwell plate coated with anti-NGAL (rat) antibody (96 wells), 5X sample diluent, NGAL rat calibrators 1 - 8, 25X wash solution concentrate, biotinylated rat NGAL antibody, HRP-streptavidin, TMB substrate and stop solution.