High-capacity oxygen trap
Supplier: Restek
20601 20600
REST20601EA 264 EUR
REST20601 REST20600
High-capacity oxygen trap
Filters Gas Filters
These high-capacity oxygen trap removes up to 135 ml of oxygen or 2 g of water.
  • Long life, typically purifies more than 480 cu ft of inert gas
  • Reduces oxygen to 15 ppb
Maximum operating pressure: 250 psi
Maximum flow: 3 L/min
Dimensions: 279×38 mm