Sterile transfer caps for Erlenmeyer flasks, Corning®
Supplier: Corning
431444 431445 431446 431447 431448 431449
734-4209EA 574.34 EUR
734-4209 734-4210 734-4211 734-4212 734-4213 734-4214
Sterile transfer caps for Erlenmeyer flasks, Corning®
Closures Safety Caps and Safety Waste Caps
Transfer caps with two ports. One port ends in a 50 mm, 0,2 μm filter disc and the other port is C-Flex Tubing ending in either a male Luer lock or a male polycarbonate quick connect.
  • Diptube reaches all the way to the bottom of the flask for easy aseptic transfer of liquid
  • Suitable for 1, 2 and 3 litre plastic Erlenmeyer flasks
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904