Guard/retention gap columns, hydroguard® water-resistant (fused silica)
Supplier: Restek
10076 10075 10077 10078 10079 10080 10081 10082 10083 10084 10085 10079-600 10080-600 10081-600 10084-600 10085-600 10086 10087 10090
REST10076EA 148 EUR
REST10076 REST10075 REST10077 REST10078 REST10079 REST10080 REST10081 REST10082 REST10083 REST10084 REST10085 REST10079-600 REST10080-600 REST10081-600 REST10084-600 REST10085-600 REST10086 REST10087 REST10090
Guard/retention gap columns, hydroguard® water-resistant (fused silica)
Chromatography Columns
Extend analytical column lifetime by preventing degradation from harsh 'steam-cleaning' water injections.
  • Tested with a comprehensive test mix, to ensure high inertness
Maximum temperature: 325 °C