Ferrule removal kit
Supplier: Restek
REST20146EA 85 EUR
REST20146
Ferrule removal kit
Puller and Inserter Tools
The tapered tools in this kit have teeth designed to grip and remove fused silica adapter ferrules that have become stuck in the fitting detail.
Delivery information: Each kit contains two tools: one for removing 0,8 mm adapter ferrules and one for removing 1,6 mm adapter ferrules.