Internal union, zero dead volume, Valco®
Supplier: Restek
Valco®
REST20147EA 112 EUR
REST20147 REST20148 REST20149 REST20150
Internal union, zero dead volume, Valco®
Fittings and Connectors
Internal uniona are made of 300 series stainless steel with end of tubing that seats squarely at bottoms of fitting details. Suitable for 1,6 mm OD tubing.
Delivery information: Supplied with stainless steel ferrules.