Internal adapter, male pipe to Valco®
Supplier: Restek
20158 20159
REST20158EA 57 EUR
REST20158 REST20159
Internal adapter, male pipe to Valco®
Fittings and Connectors
Internal adapter made of stainless steel gives minimum volume connection from a female pipe fitting on a pressure gauge or regulator to a Valco® zero-dead-volume fitting.
Delivery information: Supplied with stainless steel ferrules.