Cuvettes for fluorescence measurements
Supplier: SARSTEDT
67.754 67.741 67.755 67.738 67.745
SARS67.754EA 322 EUR
SARS67.754 SARS67.741 SARS67.755 SARS67.738 SARS67.745
Cuvettes for fluorescence measurements
Cuvettes Spectrophotometer Cuvettes
Semi-micro cuvettes in acrylic (PMMA) or polystyrene (PS).