Inlet liner packing tool
Supplier: Restek
REST20339EA 118 EUR
REST20339
Inlet liner packing tool
Puller and Inserter Tools
The inlet liner packing tool can be used with all different liners.
  • Accurate to a specific measured depth
  • Position wool reproducibly every time