Capillary installation gauge for Agilent 5973/5975 MS
Supplier: Restek
REST21894EA 265 EUR
REST21894
Capillary installation gauge for Agilent 5973/5975 MS
Puller and Inserter Tools
Capillary installation gauge made from stainless steel seats ferrules onto column for consistent installations.