Miniature passive air samplers
Supplier: Restek
26252 26253 26254 26255 26256 26257 26258 26259
REST26252EA 1420 EUR
REST26252 REST26253 REST26254 REST26255 REST26256 REST26257 REST26258 REST26259
Miniature passive air samplers
Samplers Air Samplers
These air samplers provide accurate integrated sampling via small fittings that easily connect to miniature canisters.
Informace pro objednávání: Miniature canisters are not included and must be order separately.
Delivery information: Includes a flow controller, critical orifice, vaccum gauge, sample inlet, and frit filter.