FID jet removal tool
Supplier: Restek
REST22328EA 68 EUR
REST22328
FID jet removal tool
Puller and Inserter Tools
These FID jet removal tool for Agilent 5890/6850/6890/7890 FID.
  • Securely grips jet in socket for easy removal or installation
  • Unique, ergonomic handle and easy to hold