Gas traps and connector kits, Click-On Inline Super Clean®
Supplier: Restek
22458 22460
REST22458EA 550 EUR
REST22458 REST22460
Gas traps and connector kits, Click-On Inline Super Clean®
This gas purification system with triple gas trap is ideal for purifying carrier gas and it contains oxygen, moisture, and hydrocarbon scrubbers in one cartridge.
  • Click-On fittings for easy, leak-tight cartridge changes
  • High-purity output ensures 99.9999% pure gas
Click-On adaptor connectors allow cartridges to be exchanged without introducing oxygen, moisture and hydrocarbons. Spring-loaded check valves seal when a filter is removed and open only when a new filter has been locked in place.