Septum nut removal tool
Supplier: Restek
REST24918EA 134 EUR
REST24918
Septum nut removal tool
Puller and Inserter Tools
This septum nut removal tool is ideal for Agilent 5890/6850/6890/7890 GCs and it easily removes the septum nut without touching the heated nut.
  • Unique, ergonomic handle and easy to grip
  • Nut remains in tool for quick reattachment