Piston seal insertion tool, HPLC pumps
Supplier: Restek
REST21356EA 83 EUR
REST21356
Piston seal insertion tool, HPLC pumps
Puller and Inserter Tools
Piston seal insertion tool is an alternative for paper clip and screw to remove the worn seals from the HPLC pump.
  • Simplify pump maintenance
  • Removes old piston seal and installs new seal
  • It cannot mar the surrounding metal surface of the pump housing