Solvent filter, bluestem glass
Supplier: Restek
REST26431EA 50 EUR
REST26431
Solvent filter, bluestem glass
Filters Liquid Process Filters
This solvent filter is made up of borosilicate glass and is used to prevent the particulates and microbial growth in the solvents from entering the system.
  • Provides clean mobile phase to extend the life of columns and pump seals
  • Blue filter stem allows instant visual confirmation of upright filter orientation
  • Connects to standard Ext.Ø 3,2 mm PTFE tubing using frit adaptor
  • Borosilicate frit sits lower than conventional glass filters to draw more mobile phase from each bottle
Delivery information: Includes adaptors.