Silicone oil, ~100 mPa.s AS 100, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Polyphenyl-methylsiloxane
10838-100ML
10838-500ML
SIAL10838-100MLEA
79.9
EUR
SIAL10838-100ML
SIAL10838-500ML
Silicone oil, ~100 mPa.s AS 100, Sigma-Aldrich®
Silicone oil
Silicone oil AS 100 is a viscous liquid used to study the compatibility of segmenting fluids for two phase flow applications in biomedical microdevices.
Density:
1.06 g/ml |
MDL Number:
MFCD00132673 CAS Number: 63148-58-3 |
Specification Test Results
Appearance (Color) | Colorless |
Appearance (Form) | Liquid or Viscous Liquid |
Refractive Index N20/D | 1,420 - 1,424 |
Viscosity (Rotation) | 90 - 110 mPa |
Viscosity (Conditions) | Neat, 25 °C |
Stability | Especially good thermostability (−60 - +200 °C) |