Contact Plates with Locking-Lid Plate
Supplier: PHARMAMEDIA DR MULLER
100.0100 120.0100 118.0100
MULL100.0100EA 312 EUR
MULL100.0100 MULL120.0100 MULL118.0100
Contact Plates with Locking-Lid Plate
Media for Microbiology Contact Plates for Microbiology
Used in pharmaceutical industry in clean rooms and isolators for contact sampling, personnel monitoring, as well as active air monitoring. Isolation and growth of fastidious bacteria, yeasts and moulds incubations in vent and closed position possible. Easy handling due to increased handling area.
  • Detection of aerobic and anaerobic micro-organisms
  • Gamma irradiated and triple-bagged
  • Locking-lid plate, made from polystyrene guarantees save transport
  • Label on side of bottom
  • High barrier foil for H₂O₂ as well as for water vapour
Certifikace: According Ph. Eur. and Pharmacopoeia.
Delivery information: All plates are delivered in the non-locked position, and will be delivered refrigerated.
Balení: 10 plates per packaging (6 or 10 staples) unit.