VWR® VisiCam® TV Analog Camera
Supplier: VWR
VWR® VisiCam®
630-1120EA 255 EUR
630-1120
VWR® VisiCam® TV Analog Camera
Cameras
Analog camera that can be connected to video projectors, external monitors or a computer via the optional USB set.
  • ¹/₃" CCD sensor (798×548 pixels)
  • White balance: automatic/manual
  • Resolution: 470 TV lines
Informace pro objednávání: Storage and analysis of images via the optional USB converter kit, which needs to be ordered separately.
Delivery information: Supplied with an eyepiece adapter, a CS-C mount adapter, a calibration slideand CCTV lens.