Electrode Adapter Cables
Supplier: Thermo Fisher Scientific
Orion™
ORIO91CLM0EA 269 EUR
ORIO91CLM0 662-1535 662-0699 ORIO91USRF 662-0697
Electrode Adapter Cables
Cables
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.