J.T.Baker® BAKERBOND spe™ spe-500™, SPE Columns
Supplier: Avantor
J.T.Baker® BAKERBOND spe™
7222-06EA 211 EUR
7222-06
J.T.Baker® BAKERBOND spe™ spe-500™, SPE Columns
Chromatography Columns
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum or positive pressure
General applications: Organichlorine pesticides from water.
These SPE columns can be used in reversed phase mode.
EPA approved for organochloropesticides.