Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BOSSBS-4622R-HRPEA 842 EUR
BOSSBS-4622R-HRP
Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Mycobacterium bovis

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Mycobacterium bovis
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR