Reusable half masks, X-plore® 3300 and X-plore® 3500
Supplier: Dräger
R55330 R55331 R55332 R55350 R55351 R55352 R56960
111-0416EA 24.05 EUR
111-0416 111-0415 111-0414 111-0413 111-0412 111-0411 111-0407
Reusable half masks, X-plore® 3300 and X-plore® 3500
Respirators Reusable Respirators Half Face Respirators
The X-plore® 3300 and 3500 half masks are low maintenance and affordable. They are equipped with two lateral bayonet connections that enable a whole range of bayonet filters to be used.
  • Flexible nose section provides an extra tight seal and excellent fit even when wearing safety goggles
  • Side filters and a shallow design, give an exceptional field of vision and X-plore® fits perfectly under protective visors
  • The soft FlexiFit head cradle is a perfect fit for all head shapes - without trapping hair; it is also barely noticeable under helmets
  • Easily adjustable head strap
X-plore 3500 - special features:
The mask body is made from an innovative, new material: Very kind to the skin, soft and silicone-free; features that combine premium comfort with excellent durability
Drop-down strap option: By unclasping the neck strap, the mask can be removed from your face without taking off your head protection; the inner surface of the mask faces the body, protecting it from contamination
Weight: 95 g
Certifikace: EN 140