Fat test strips, (LRSM)
Supplier: 3M
MMMAFETT200EA 403 EUR
MMMAFETT200
Fat test strips, (LRSM)
Food and Beverage Test Systems
The fat test strip is to determine fat quality by measuring the free fatty acids.
  • Operation at temperatures up to 180 °C
  • Easy and fast solution method
This rapid test is suitable for use with the same fat and the same fried food. The result can be easily read in approx. 15 to 30 seconds, by means of a colour change.