ESD safety ankle boots, lace-up, SL 845 XP blue
Supplier: ATLAS - the shoe company
65200_W10_36 65200_W10_37 65200_W10_38 65200_W10_39 65200_W10_40 65200_W10_41 65200_W10_42 65200_W10_43 65200_W10_44 65200_W10_45 65200_W10_46 65200_W10_47 65200_W10_48 65200_W10_49 65200_W12_36 65200_W12_37 65200_W12_38 65200_W12_39 65200_W12_40 65200_W12_41 65200_W12_42 65200_W12_43 65200_W12_44 65200_W12_45 65200_W12_46 65200_W12_47 65200_W12_48 65200_W12_49 65200_W13_36 65200_W13_37 65200_W13_38 65200_W13_39 65200_W13_40 65200_W13_41 65200_W13_42 65200_W13_43 65200_W13_44 65200_W13_45 65200_W13_46 65200_W13_47 65200_W14_36 65200_W14_37 65200_W14_38 65200_W14_39 65200_W14_40 65200_W14_41 65200_W14_42 65200_W14_43 65200_W14_44 65200_W14_45 65200_W14_46 65200_W14_47 65200_W14_48 65200_W14_49 65200_W13_48 65200_W13_49
114-7942EA 248 EUR
114-7942 114-7943 114-7944 114-7945 114-7946 114-7947 114-7948 114-7949 114-7950 114-7951 114-7952 114-7953 114-7954 114-7955 114-7956 114-7957 114-7958 114-7959 114-7960 114-7961 114-7962 114-7963 114-7964 114-7965 114-7966 114-7967 114-7968 114-7969 114-7913 114-7914 114-7915 114-7916 114-7917 114-7918 114-7919 114-7920 114-7921 114-7922 114-7923 114-7924 114-7927 114-7928 114-7929 114-7930 114-7931 114-7932 114-7933 114-7934 114-7935 114-7936 114-7937 114-7938 114-7939 114-7940 114-7925 114-7926
ESD safety ankle boots, lace-up, SL 845 XP blue
Shoes and Boots
ESD safety ankle boots made from waterproof leather upper, light lining materials and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Puncture resistant, metal-free XP® midsole
  • Lightweight alu-tec® toecap made from aluminium (200 J)
  • Carbon coated toe protection
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Black/blue
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3