Anti-Biotin Goat Polyclonal Antibody (DyLight® 350)

Supplier: Immunoreagents
IMMRIR2962EA 224 EUR
IMMRIR2962
Anti-Biotin Goat Polyclonal Antibody (DyLight® 350)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Turkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR