Methyl ethyl ketone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
5447-25EA 118.55 EUR
5447-25
Methyl ethyl ketone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72,11 g/mol
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://si.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg