n-Hexane, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
Synonyms: Hexyl hydride, Hexanen, Hexane
6729-25EA 161.5 EUR
6729-25 6729-45
n-Hexane, AR®, Laboratory Reagent, Macron Fine Chemicals™
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM