Sodium hydroxide 97.0-100.5% Ph. Eur., Macron Fine Chemicals™
Supplier: Avantor Performance Materials
Synonyms: Caustic soda
7066-33EA 30.19 EUR
7066-33 7066-36 7066-38
Sodium hydroxide 97.0-100.5% Ph. Eur., Macron Fine Chemicals™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II