Crucibles, quartz, self sealing
Supplier: CEM
458-4221EA 238 EUR
458-4221
Crucibles, quartz, self sealing
Crucibles
This quartz crucible is self sealing and enables oxygen free ashing.
Delivery information: Includes crucible and lid.