Surgical scalpel handle, SF13
Supplier: Swann-Morton
SWAN6061EA 99.9 EUR
SWAN6061
Surgical scalpel handle, SF13
Scalpels and Scalpel Assemblies Scalpel Handles
Stainless steel.
Fits blades SM61, SM62, SM63, SM64, SM65, SM65A, SM67, SM68, SM69, SP90, SP91 and Myringotomy blade.
Balení: Individually sleeved.