ESD safety rigger boots, Safety-Grip, pull-on
Supplier: ELTEN
78671/38 78671/39 78671/40 78671/41 78671/42 78671/43 78671/44 78671/45 78671/46 78671/47 78671/48 78671/49 78671/50 78671/36 78671/37
114-8227EA 105.9 EUR
114-8227 114-8228 114-8229 114-8230 114-8231 114-8232 114-8233 114-8234 114-8235 114-8236 114-8237 114-8238 114-8239 114-8225 114-8226
ESD safety rigger boots, Safety-Grip, pull-on
Shoes and Boots
These ESD winter safety boots are made from cowhide leather with a PU/PU walking sole (Safety-Grip).
  • Warm lining
  • Full length warm fleece, ESD inlay sole
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Brown
Certifikace: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC, CI.