ESD safety shoes, Velcro® fastening, SL 26 Green
Supplier: ATLAS - the shoe company
232/35 232/36 232/37 232/38 232/39 232/40 232/41 232/42 232/43 232/44 232/45 232/46 232/47 232/48 232/49
ATLA232/35EA 0 EUR
ATLA232/35 ATLA232/36 ATLA232/37 ATLA232/38 ATLA232/39 ATLA232/40 ATLA232/41 ATLA232/42 ATLA232/43 ATLA232/44 ATLA232/45 ATLA232/46 ATLA232/47 ATLA232/48 ATLA232/49
ESD safety shoes, Velcro® fastening, SL 26 Green
Shoes and Boots
Lightweight ESD safety shoe made from Sportline upper (oiled nubuck leather), breathable functional lining and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Lightweight alu-tec® toecap made from aluminium (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/green
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC