ESD safety shoes, Velcro® fastening, SL 46 Blue
Supplier: ATLAS - the shoe company
233/35 233/36 233/37 233/38 233/39 233/40 233/41 233/42 233/43 233/44 233/45 233/46 233/47 233/48 233/49
ATLA233/35EA 0 EUR
ATLA233/35 ATLA233/36 ATLA233/37 ATLA233/38 ATLA233/39 ATLA233/40 ATLA233/41 ATLA233/42 ATLA233/43 ATLA233/44 ATLA233/45 ATLA233/46 ATLA233/47 ATLA233/48 ATLA233/49
ESD safety shoes, Velcro® fastening, SL 46 Blue
Shoes and Boots
Lightweight ESD safety shoe made from Sportline upper (oiled nubuck leather), breathable functional lining and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure-relieving insole, cushioning comfort midsole made from MPU, dynamic outsole system
  • Lightweight alu-tec® toecap made from aluminium (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/blue
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC