Preservative, ProClin™ 300, Sigma-Aldrich®
Supplier: Merck
48912-U 48914-U 48915-U 48917-U 48918-U
SIAL48912-UEA 93.3 EUR
SIAL48912-U SIAL48914-U SIAL48915-U SIAL48917-U SIAL48918-U
Preservative, ProClin™ 300, Sigma-Aldrich®
Cleaning Agents Disinfectants
ProClin™ are among the most widely used presevatives in the in vitro diagnostic (IVD) industry. These materials are water-soluble formulations that are easily used directly from the container. They have no health hazards, toxicology problems, or disposal issues at recommended usage levels.
  • Compatibility with key enzymes and diagnostic assay indicators
  • Will not inhibit antibody binding
  • Excellent stability over a wide pH range
  • Stable in TRIS, TES and HEPES buffer up to a pH of 7
  • Level of active ingredients assayable by HPLC
  • Safe and easy to use, water soluble liquid
ProClin™ presevatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin™ toxicity prevent microorganisms from developing high levels of resistance, making ProClin™ highly efficient even over extended storage periods of your IVD components.
Appearance: Clear, colorless to light yellow liquid, free of foreign matter.